missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tribehenin, TCI America™

Manufacturer:  TCI America T13915G

 View more versions of this product

Catalog No. T13915G



Specifications

Specifications

Tribehenin
83°C
18641-57-1
MFCD00056274
DMBUODUULYCPAK-UHFFFAOYSA-N
2,3-di(docosanoyloxy)propyl docosanoate
62726
Crystal-Powder at 20°C
White
5g
C69H134O6
Glycerol Tridocosanoate, 1,2,3-Tridocosanoyl Glycerol
CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC
1059.825
1059.83
≥90.0% (T)

Chemical Identifiers

18641-57-1
1059.825
DMBUODUULYCPAK-UHFFFAOYSA-N
62726
CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC
C69H134O6
MFCD00056274
Glycerol Tridocosanoate, 1,2,3-Tridocosanoyl Glycerol
2,3-di(docosanoyloxy)propyl docosanoate
Safety and Handling

Safety and Handling

RTECSNumber : JR1305000

TSCA : No

Documents