missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triallylamine, 98%

Manufacturer:  Alfa Aesar™ A1208130

 View more versions of this product

Catalog No. AAA1208130



Description

Description

Applications
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from strong oxidizing agents and acids. Protect against electrostatic charges and keep away from ignition sources.
Specifications

Specifications

Triallylamine
Amine-like
-70°C
102-70-5
MFCD00026093
triallylamine, tris 2-propenyl amine, 2-propen-1-amine, n,n-di-2-propenyl, triallyl amine, unii-b6n19xc04r, ccris 4876, n,n-diallylprop-2-en-1-amine, n,n-di-2-propenyl-2-propen-1-amine, ch2=chch2 3n, 4-04-00-01061 beilstein handbook reference
C=CCN(CC=C)CC=C
137.226
137.23
31°C (87°F)
150°C to 151°C
250g
C9H15N
UN2610
VPYJNCGUESNPMV-UHFFFAOYSA-N
N,N-bis(prop-2-enyl)prop-2-en-1-amine
7617
98%

Chemical Identifiers

102-70-5
137.226
VPYJNCGUESNPMV-UHFFFAOYSA-N
7617
C=CCN(CC=C)CC=C
C9H15N
MFCD00026093
triallylamine, tris 2-propenyl amine, 2-propen-1-amine, n,n-di-2-propenyl, triallyl amine, unii-b6n19xc04r, ccris 4876, n,n-diallylprop-2-en-1-amine, n,n-di-2-propenyl-2-propen-1-amine, ch2=chch2 3n, 4-04-00-01061 beilstein handbook reference
N,N-bis(prop-2-enyl)prop-2-en-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H311-H314-H318-H226-H302-H332
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIALLYLAMINE

RTECSNumber : XX5950000

TSCA : TSCA

SDS Documents