Please login to your online account to display your discounted pricing

Alfa Aesar™ Triallyl phosphite

Chemical Identifiers

Chemical Name or Material Triallyl phosphite
CAS 102-84-1
Linear Formula (CH2=CHCH2O)3P
Formula Weight 202.19
MDL Number MFCD00014948
UN Number UN3278
Beilstein 1705535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1748409

 
alfa aesar™
1748409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1748418

 
alfa aesar™
1748418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1748430

 
alfa aesar™
1748430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.19
Chemical Name or Material Triallyl phosphite
TSCA TSCA
Physical Form Liquid
Linear Formula (CH2=CHCH2O)3P
MDL Number MFCD00014948
CAS 102-84-1
UN Number UN3278
Beilstein 1705535
Color Colorless
Boiling Point 84°C (12mmHg)
Health Hazard 1 Danger
Quantity 10g
Flash Point 86° (187°F)
Odor Strong
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
RTECS Number TH1772500
Safety and Handling

Danger

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.