missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triacetylresveratrol 98.0+%, TCI America™
$28.21 - $84.60
Chemical Identifiers
CAS | 42206-94-0 |
---|---|
Molecular Formula | C20H18O6 |
Molecular Weight (g/mol) | 354.358 |
MDL Number | MFCD01546481 |
InChI Key | PDAYUJSOJIMKIS-SNAWJCMRSA-N |
PubChem CID | 5962587 |
IUPAC Name | [4-[(E)-2-(3,5-diacetyloxyphenyl)ethenyl]phenyl] acetate |
SMILES | CC(=O)OC1=CC=C(C=C1)C=CC2=CC(=CC(=C2)OC(=O)C)OC(=O)C |
Chemical Identifiers
42206-94-0 | |
354.358 | |
PDAYUJSOJIMKIS-SNAWJCMRSA-N | |
[4-[(E)-2-(3,5-diacetyloxyphenyl)ethenyl]phenyl] acetate |
C20H18O6 | |
MFCD01546481 | |
5962587 | |
CC(=O)OC1=CC=C(C=C1)C=CC2=CC(=CC(=C2)OC(=O)C)OC(=O)C |
Specifications
42206-94-0 | |
White | |
MFCD01546481 | |
3,4′C,5-Triacetoxy-trans-stilbene | |
CC(=O)OC1=CC=C(C=C1)C=CC2=CC(=CC(=C2)OC(=O)C)OC(=O)C | |
354.358 | |
354.36 | |
Crystalline Powder |
119°C | |
C20H18O6 | |
1 g | |
PDAYUJSOJIMKIS-SNAWJCMRSA-N | |
[4-[(E)-2-(3,5-diacetyloxyphenyl)ethenyl]phenyl] acetate | |
5962587 | |
≥98.0% (GC) | |
Triacetylresveratrol |
Safety and Handling
TSCA : No