missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triacetoxyethylsilane, TCI America™

Manufacturer:  TCI America E0525500G

 View more versions of this product

Catalog No. E0525500G



Specifications

Specifications

Triacetoxyethylsilane
Yellow
17689-77-9
MFCD00053703
ethyltriacetoxysilane, triacetoxyethylsilane, triacetoxy ethyl silane, triacetoxy ethylsilane, ethylsilanetriyl triacetate, silanetriol, ethyl-, triacetate, silanetriol, 1-ethyl-, 1,1,1-triacetate, dsstox_cid_7792, dsstox_rid_78567, dsstox_gsid_27792
CC[Si](OC(=O)C)(OC(=O)C)OC(=O)C
234.279
234.28
≥96.0% (GC)
109°C
500g
C8H14O6Si
1760
KXJLGCBCRCSXQF-UHFFFAOYSA-N
[diacetyloxy(ethyl)silyl] acetate
87244
Clear Liquid at 20°C

Chemical Identifiers

17689-77-9
234.279
KXJLGCBCRCSXQF-UHFFFAOYSA-N
87244
CC[Si](OC(=O)C)(OC(=O)C)OC(=O)C
C8H14O6Si
MFCD00053703
ethyltriacetoxysilane, triacetoxyethylsilane, triacetoxy ethyl silane, triacetoxy ethylsilane, ethylsilanetriyl triacetate, silanetriol, ethyl-, triacetate, silanetriol, 1-ethyl-, 1,1,1-triacetate, dsstox_cid_7792, dsstox_rid_78567, dsstox_gsid_27792
[diacetyloxy(ethyl)silyl] acetate
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1939

TSCA : Yes

Documents