Please login to your online account to display your discounted pricing

Alfa Aesar™ Triacetoxy(vinyl)silane, 95%

Chemical Identifiers

Chemical Name or Material Triacetoxy(vinyl)silane
CAS 4130-08-9
Molecular Formula C8H12O6Si
Formula Weight 232.26
MDL Number MFCD00014975
UN Number UN1760
Beilstein 1792454
Synonym Vinyltriacetoxysilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB242120B

  SDS
alfa aesar™
B242120B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2421218

  SDS
alfa aesar™
B2421218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2421230

  SDS
alfa aesar™
B2421230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.26
Chemical Name or Material Triacetoxy(vinyl)silane
TSCA TSCA
Assay 95%
Molecular Formula C8H12O6Si
MDL Number MFCD00014975
CAS 4130-08-9
UN Number UN1760
Beilstein 1792454
Synonym Vinyltriacetoxysilane
Boiling Point 112° to 113°C (1mmHg)
Melting Point 7°C
Quantity 1000g
Flash Point 76°C (168°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

Applications
Triacetoxy(vinyl)silane is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.