missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triacetin, USP, 97-102%, Spectrum™ Chemical
TR106, 102-76-1, C9H14O6
$1714.17 - $1714.17
Chemical Identifiers
CAS | 102-76-1 |
---|---|
Molecular Formula | C9H14O6 |
Molecular Weight (g/mol) | 218.21 |
InChI Key | URAYPUMNDPQOKB-UHFFFAOYSA-N |
IUPAC Name | 1,3-bis(acetyloxy)propan-2-yl acetate |
SMILES | CC(=O)OCC(COC(C)=O)OC(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18607507
|
Spectrum Chemical Mfg Cor
TR1064LTGL |
4 L | Amber Glass Bottle |
Each for $1,714.17
|
|
||||
Description
Spectrum™ Chemical Triacetin, USP is used as an antifungal agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
102-76-1 | |
218.21 | |
1,3-bis(acetyloxy)propan-2-yl acetate |
C9H14O6 | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
CC(=O)OCC(COC(C)=O)OC(C)=O |
Specifications
102-76-1 | |
Amber Glass Bottle | |
4 L | |
CC(=O)OCC(COC(C)=O)OC(C)=O | |
218.21 | |
USP |
1 | |
C9H14O6 | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
1,3-bis(acetyloxy)propan-2-yl acetate | |
97 to 102% |