missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triacetin, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139220100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triacetin | |
3.0°C | |
1.1550g/mL | |
138°C | |
98.5% min. (GC) | |
C9H14O6 | |
(CH3CO2CH2)2CH(O2CCH3) | |
02,147 | |
15,9751 | |
Solubility in water: 64.0g/L (20°C). Other solubilities: soluble in most common organic solvents,slightly soluble in carbon disulfide | |
CC(=O)OCC(COC(=O)C)OC(=O)C | |
218.21 | |
23 mPa.s (20°C) | |
218.21 | |
Liquid |
102-76-1 | |
Colorless | |
258.0°C | |
Authentic | |
Plastic drum | |
1.4290 to 1.4330 | |
10 L | |
1.155 | |
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
2,3-diacetyloxypropyl acetate | |
5541 | |
CHEBI:9661 | |
99% |
Chemical Identifiers
102-76-1 | |
218.21 | |
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine | |
CHEBI:9661 | |
CC(=O)OCC(COC(=O)C)OC(=O)C |
C9H14O6 | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
5541 | |
2,3-diacetyloxypropyl acetate |
Safety and Handling
EINECSNumber : 203-051-9
RUO – Research Use Only