Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-tert-butylphosphonium tetrafluoroborate, 97%

$120.69 - $386.36

Chemical Identifiers

Chemical Name or Material Tri-tert-butylphosphonium tetrafluoroborate
CAS 131274-22-1
Molecular Formula C12H28BF4P
Formula Weight 290.13
MDL Number MFCD07189501
Beilstein 8813613
Synonym Fluoroboric acid tri-tert-butylphosphine adduct; Tri-tert-butylphosphine fluoroboric acid adduct
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1975203

 
alfa aesar™
L1975203
1g Each for $120.69

AAL1975206

 
alfa aesar™
L1975206
5g Each for $386.36
Description & Specifications

Specifications

Formula Weight 290.13
Chemical Name or Material Tri-tert-butylphosphonium tetrafluoroborate
Assay 97%
MDL Number MFCD07189501
Synonym Fluoroboric acid tri-tert-butylphosphine adduct; Tri-tert-butylphosphine fluoroboric acid adduct
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C12H28BF4P
CAS 131274-22-1
Beilstein 8813613
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 266° to 267°C
Quantity 1g

Applications
Tri-tert-butylphosphonium tetrafluoroborate is a ligand used in the palladium-catalyzed enantioselective alfa-arylation of N-boc-pyrrolidine. It is also used with a palladium(0)-15-membered, triolefinic, macrocyle in Suzuki cross-coupling reactions of aryl bromides and chlorides. Further, it is used in the Heck coupling of vinyl tosylates with olefins.

Solubility
Soluble in methylene chloride and chloroform. Slightly soluble in terahydro furan. Insoluble in hexane, toluene and water.

Notes
Incompatible with strong oxidizing agents and halogens.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.