missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetate, TCI America™

Manufacturer:  TCI America T26421G

 View more versions of this product

Catalog No. T26421G



Specifications

Specifications

Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetate
1g
C28H52N4O8
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid Tri-tert-butyl Ester
CC(C)(C)OC(=O)CN1CCN(CCN(CCN(CC1)CC(=O)OC(C)(C)C)CC(=O)OC(C)(C)C)CC(=O)O
572.744
572.74
≥97.0% (T)
Undesignated
137076-54-1
MFCD02259697
RVUXZXMKYMSWOM-UHFFFAOYSA-N
2-[4,7,10-tris[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
11606627
Crystal-Powder at 20°C

Chemical Identifiers

137076-54-1
572.744
RVUXZXMKYMSWOM-UHFFFAOYSA-N
11606627
CC(C)(C)OC(=O)CN1CCN(CCN(CCN(CC1)CC(=O)OC(C)(C)C)CC(=O)OC(C)(C)C)CC(=O)O
C28H52N4O8
MFCD02259697
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid Tri-tert-butyl Ester
2-[4,7,10-tris[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents