Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri(p-tolyl)phosphine, 98%

$23.54 - $321.72

Chemical Identifiers

Chemical Name or Material Tri(p-tolyl)phosphine
CAS 1038-95-5
Molecular Formula C21H21P
Formula Weight 304.37
MDL Number MFCD00008542
Beilstein 651045
Synonym Tris(4-methylphenyl)phosphine; Tris(p-tolyl)phosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1402103

  SDS
alfa aesar™
A1402103
1g Each for $23.54

AAA1402106

  SDS
alfa aesar™
A1402106
5g Each for $76.65

AAA1402114

  SDS
alfa aesar™
A1402114
25g Each for $321.72
Description & Specifications

Specifications

Formula Weight 304.37
Chemical Name or Material Tri(p-tolyl)phosphine
TSCA TSCA
Assay 98%
Molecular Formula C21H21P
MDL Number MFCD00008542
CAS 1038-95-5
Beilstein 651045
Synonym Tris(4-methylphenyl)phosphine; Tris(p-tolyl)phosphine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 144° to 146°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SZ3880000

Applications
Tri(p-tolyl)phosphine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.