missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri-o-tolylbismuth Dichloride, TCI America™

Manufacturer:  TCI America T19575G

 View more versions of this product

Catalog No. T19575G



Specifications

Specifications

Tri-o-tolylbismuth Dichloride
5g
C21H21BiCl2
Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride
CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)(C3=CC=CC=C3C)(Cl)Cl
553.279
553.28
≥97.0% (T)
Undesignated
6729-60-8
MFCD00092459
RLWWKAGRZATJDC-UHFFFAOYSA-L
dichloro-tris(2-methylphenyl)bismuth
5225331
Crystal-Powder at 20°C

Chemical Identifiers

6729-60-8
553.279
RLWWKAGRZATJDC-UHFFFAOYSA-L
5225331
CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)(C3=CC=CC=C3C)(Cl)Cl
C21H21BiCl2
MFCD00092459
Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride
dichloro-tris(2-methylphenyl)bismuth
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents