missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri(o-tolyl) phosphite, typically C 71%, H 6%

Manufacturer:  Alfa Aesar™ 1025806

 View more versions of this product

Catalog No. AA1025806



Specifications

Specifications

Tri(o-tolyl) phosphite
Air Sensitive
Colorless
2632-10-2
MFCD00014911
Typically C 71%, H 6%
C1=CC(=C(C=C1C(=O)CBr)Cl)Cl
267.93
352.36
>110° (230°F)
200°C to 202°C (2.4mmHg to 2.6mmHg)
5g
C8H5BrCl2O
2-bromo-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl bromide, 2-bromo-3',4'-dichloroacetophenone, 2-bromo-1-3,4-dichlorophenyl ethan-1-one, 3,4-dichlorophenacylbromide, ethanone, 2-bromo-1-3,4-dichlorophenyl, 1-3,4-dichlorophenyl-2-bromoethan-1-one, pubchem3262, acmc-1coei, 3.4-dichlorophenacylbromide
PAKFHEFMTRCFAU-UHFFFAOYSA-N
2-bromo-1-(3,4-dichlorophenyl)ethanone
244751
Liquid

Chemical Identifiers

2632-10-2
267.93
PAKFHEFMTRCFAU-UHFFFAOYSA-N
244751
C1=CC(=C(C=C1C(=O)CBr)Cl)Cl
C8H5BrCl2O
MFCD00014911
2-bromo-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl bromide, 2-bromo-3',4'-dichloroacetophenone, 2-bromo-1-3,4-dichlorophenyl ethan-1-one, 3,4-dichlorophenacylbromide, ethanone, 2-bromo-1-3,4-dichlorophenyl, 1-3,4-dichlorophenyl-2-bromoethan-1-one, pubchem3262, acmc-1coei, 3.4-dichlorophenacylbromide
2-bromo-1-(3,4-dichlorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

RTECSNumber : TH1050000

Documents