Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri(o-tolyl) phosphite, typically C 71%, H 6%

$119.93 - $424.91

Chemical Identifiers

Chemical Name or Material Tri(o-tolyl) phosphite
CAS 2622-08-4
Formula Weight 352.36
MDL Number MFCD00014911
Synonym Tri-o-cresyl phosphite; Tris(2-tolyl)phosphite
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1025806

  SDS
alfa aesar™
1025806
5g Each for $119.93

AA1025814

  SDS
alfa aesar™
1025814
25g Each for $424.91
Description & Specifications

Specifications

Formula Weight 352.36
Color Colorless
Sensitivity Air Sensitive
MDL Number MFCD00014911
Quantity 5g
Physical Form Liquid
Flash Point >110° (230°F)
Chemical Name or Material Tri(o-tolyl) phosphite
Concentration Typically C 71%, H 6%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 200° to 202°C (2.4mmHg to 2.6mmHg)
CAS 2622-08-4
Synonym Tri-o-cresyl phosphite; Tris(2-tolyl)phosphite
RTECS Number TH1050000
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.