Please login to your online account to display your discounted pricing

Tri-o-tolyl phosphate, 96%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC366461000

 View more versions of this product

Catalog No. AC366461000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Viscous Liquid
Formula Weight 368.36
Quantity 100g
Color Clear colorless to yellow
Merck Index 15, 9941
Density 1.2g/mL
Name Note 96%
Molecular Formula C21H21O4P
Chemical Name or Material Tri-o-tolyl phosphate
MDL Number MFCD00041906
Assay Percent Range 96%
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes damage to organs.
Packaging Glass bottle
Refractive Index 1.5580 to 1.5610 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
CAS 78-30-8
Health Hazard 3 GHS P Statement:
IF exposed: Call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tri-o-tolyl phosphate
Name Note 96%
CAS 78-30-8
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C21H21O4P
Formula Weight 368.36
MDL Number MFCD00041906
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF exposed: Call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes damage to organs.

SDS & Safety Documents