missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tri-O-acetyl-D-glucal, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 139241000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Tri-O-acetyl-D-glucal | |
2873-29-2 | |
0.5% max. (30°C, 15 hrs) (vacuum) | |
98.5% min. (GC) | |
C12H16O7 | |
100g | |
−15° (20°C c=2,EtOH) | |
tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal | |
LLPWGHLVUPBSLP-UTUOFQBUSA-N | |
[(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate | |
688303 | |
99% |
99% | |
>110°C | |
Authentic | |
Glass bottle | |
MFCD00063253 | |
17, I, 111 | |
− 15.00 | |
Solubility in water: slightly soluble | |
CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C | |
272.25 | |
272.25 |
Safety and Handling
EINECSNumber : 220-709-0
RUO â Research Use Only