missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-propyltin chloride

Manufacturer:  Alfa Aesar™ 7112214

 View more versions of this product

Catalog No. AA7112214



Specifications

Specifications

Tri-n-propyltin chloride
-23.5°C
25g
1.4910
C9H21ClSn
UN2788
FVFLIQMADUVDSP-UHFFFAOYSA-M
chloro(tripropyl)stannane
16784
Liquid
123°C (13mmHg)
2279-76-7
1.2678g/mL at 28°C
Moisture Sensitive
MFCD00013610
tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride
CCC[Sn](CCC)(CCC)Cl
283.427
283.41

Chemical Identifiers

2279-76-7
283.427
FVFLIQMADUVDSP-UHFFFAOYSA-M
16784
CCC[Sn](CCC)(CCC)Cl
C9H21ClSn
MFCD00013610
tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride
chloro(tripropyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

SDS Documents