Please login to your online account to display your discounted pricing

Tri-n-propylsilane 99% , ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tri-n-propylsilane
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H22Si
Linear Formula HSi(CH2CH2CH3)3
Formula Weight 158.36
MDL Number MFCD00009358
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390320050

  SDS
thermo fisher scientific
AC390320050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390320250

  SDS
thermo fisher scientific
AC390320250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 158.36
Quantity 5g
Color Clear colorless
Density 0.758g/mL
Assay Percent Range 99%
Linear Formula HSi(CH2CH2CH3)3
Refractive Index 1.4270 to 1.4290 (20°C, 589nm)
Chemical Name or Material Tri-n-propylsilane
Infrared Spectrum Authentic
Molecular Formula C9H22Si
Packaging Glass bottle
MDL Number MFCD00009358
CAS 998-29-8
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Reduction reagent

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.