Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-propylphosphine, 98%

Chemical Identifiers

Chemical Name or Material Tri-n-propylphosphine
CAS 2234-97-1
Molecular Formula (CH3CH2CH2)3P
Formula Weight 160.24g/mol
MDL Number MFCD00015217
UN Number UN2845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2783814

 
alfa aesar™
H27838-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2783822

 
alfa aesar™
H27838-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 72° to 74°C (12mmHg)
Quantity 25g
Flash Point 62°C (144°F)
Chemical Name or Material Tri-n-propylphosphine
CAS 2234-97-1
Assay 98%
Molecular Formula (CH3CH2CH2)3P
Formula Weight 160.24g/mol
MDL Number MFCD00015217
UN Number UN2845
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H250-H252-H314-H318-H227
Catches fire spontaneously if exposed to air.
Self-heating in large quantities; may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.