missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-propylamine, 98%

Manufacturer:  Alfa Aesar™ A10285AK

 View more versions of this product

Catalog No. AAA10285AK



Description

Description

Applications
Tri-n-propylamine is used as an chemical intermediate, organic solvent. It is used in the preparation of chemical compounds such as dyes, drugs, catalyst & other uses. It is used together with ruthenium in the electrochemiluminescence.

Solubility
Soluble in water (2.6 g/L) at 20°C, ether and ethanol.

Notes
Hygroscopic. Incompatible with oxidizing agents, acids.
Specifications

Specifications

Tri-n-propylamine
Amine-like
-93°C
102-69-2
(CH3CH2CH2)3N
UN2260
YFTHZRPMJXBUME-UHFFFAOYSA-N
N,N-dipropylpropan-1-amine
7616
143.27
34°C (93°F)
156°C
250mL
C9H21N
MFCD00009363
tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
CCCN(CCC)CCC
143.274
CHEBI:38880
98%

Chemical Identifiers

102-69-2
143.274
YFTHZRPMJXBUME-UHFFFAOYSA-N
7616
N,N-dipropylpropan-1-amine
C9H21N
MFCD00009363
tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
CHEBI:38880
CCCN(CCC)CCC
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIPROPYLAMINE

RTECSNumber : TX1575000

TSCA : TSCA

SDS Documents