missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri-n-propyl orthoformate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1550306

 View more versions of this product

Catalog No. AAA1550306



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tri-n-propyl orthoformate
0.886
72°C (161°F)
1.407
5 g
Moisture sensitive
RWNXXQFJBALKAX-UHFFFAOYSA-N
1-(dipropoxymethoxy)propane
69311
97%
621-76-1
106°C to 108°C (40 mmHg)
C10H22O3
MFCD00015214
1744249
tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
CCCOC(OCCC)OCCC
190.283
190.28

Chemical Identifiers

621-76-1
190.283
RWNXXQFJBALKAX-UHFFFAOYSA-N
69311
CCCOC(OCCC)OCCC
C10H22O3
MFCD00015214
tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
1-(dipropoxymethoxy)propane
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 210-704-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only