Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-propyl borate, 98%

Chemical Identifiers

Chemical Name or Material Tri-n-propyl borate
CAS 688-71-1
Linear Formula (C3H7O)3B
Formula Weight 188.08
MDL Number MFCD00009366
UN Number UN3272
Synonym n-Propyl borate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2297430

  SDS
alfa aesar™
2297430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2297418

  SDS
alfa aesar™
2297418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA22974A1

  SDS
alfa aesar™
22974A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.08
Sensitivity Moisture Sensitive
Linear Formula (C3H7O)3B
Synonym n-Propyl borate
RTECS Number ED5785000
TSCA TSCA
CAS 688-71-1
Density 0.861g/mL
UN Number UN3272
Refractive Index 1.3950
Health Hazard 1 Warning
Color Colorless
Quantity 250g
Flash Point 32° (90°F)
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Boiling Point 175° to 177°C
MDL Number MFCD00009366
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Tri-n-propyl borate
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Solubility Information Soluble in organic solvents; Decomposes in water
Physical Form Liquid
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.