Learn More
Tri-n-propyl borate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02297430
Description
Tri-n-propyl borate is used as solvents or catalysts for the production of resins, waxes, paints and varnishes, antioxidants, flame retardants. It is also used for petroleum products, tackifiers for silicone elastomers, stabilizers and binders for rubbers or plastics and as cross-linking agents in polymerization processes. They are are used in welding fluxes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tri-n-propyl borate | |
Colorless | |
175°C to 177°C | |
C9H21BO3 | |
(C3H7O)3B | |
250 g | |
Moisture sensitive | |
Soluble in organic solvents; Decomposes in water | |
B(OCCC)(OCCC)OCCC | |
188.074 | |
188.08 | |
Liquid |
688-71-1 | |
0.861 g/mL | |
32°C (90°F) | |
1.395 | |
MFCD00009366 | |
UN3272 | |
tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester | |
LTEHWCSSIHAVOQ-UHFFFAOYSA-N | |
tripropyl borate | |
12711 | |
98% |
Chemical Identifiers
688-71-1 | |
188.074 | |
LTEHWCSSIHAVOQ-UHFFFAOYSA-N | |
12711 | |
B(OCCC)(OCCC)OCCC |
C9H21BO3 | |
MFCD00009366 | |
tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester | |
tripropyl borate |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 211-703-9
RTECSNumber : ED5785000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only