Please login to your online account to display your discounted pricing

Tri-n-hexyltrimellitate, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375555000

 View more versions of this product

Catalog No. AC375555000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 462.63
Quantity 500mL
Color Clear slightly yellow
Density 1.0150g/mL
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Linear Formula C6H3(CO2(CH2)5CH3)3
Molecular Formula C27H42O6
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material Tri-n-hexyltrimellitate
MDL Number MFCD00152931
CAS 1528-49-0

Specifications

Chemical Name or Material Tri-n-hexyltrimellitate
Assay ≥96.0%
Molecular Formula C27H42O6
Linear Formula C6H3(CO2(CH2)5CH3)3
Formula Weight 462.63
MDL Number MFCD00152931