Please login to your online account to display your discounted pricing

Tri-n-hexyltrimellitate, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tri-n-hexyltrimellitate
Assay ≥96.0%
Molecular Formula C27H42O6
Linear Formula C6H3(CO2(CH2)5CH3)3
Formula Weight 462.63
MDL Number MFCD00152931
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375555000

  SDS
thermo fisher scientific
AC375555000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 462.63
Quantity 500mL
Color Clear slightly yellow
Density 1.0150g/mL
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Linear Formula C6H3(CO2(CH2)5CH3)3
Molecular Formula C27H42O6
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material Tri-n-hexyltrimellitate
MDL Number MFCD00152931
CAS 1528-49-0