missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri-n-hexyl phosphate, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01680106

 View more versions of this product

Catalog No. AA1680106



Description

Description

Used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tri-n-hexyl phosphate
Colorless
[CH3(CH2)5O]3PO
5 g
SFENPMLASUEABX-UHFFFAOYSA-N
trihexyl phosphate
75666
≥90%
2528-39-4
C18H39O4P
MFCD00015351
phosphoric acid, trihexyl ester, tri-n-hexyl phosphate, tri n-hexyl phosphate, acmc-20aje0, aronis24109
CCCCCCOP(=O)(OCCCCCC)OCCCCCC
350.48
350.48
Liquid

Chemical Identifiers

2528-39-4
350.48
SFENPMLASUEABX-UHFFFAOYSA-N
75666
CCCCCCOP(=O)(OCCCCCC)OCCCCCC
C18H39O4P
MFCD00015351
phosphoric acid, trihexyl ester, tri-n-hexyl phosphate, tri n-hexyl phosphate, acmc-20aje0, aronis24109
trihexyl phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-774-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only