missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-hexyl phosphate, 90+%

Manufacturer:  Alfa Aesar™ 1680114

 View more versions of this product

Catalog No. AA1680114



Specifications

Specifications

Tri-n-hexyl phosphate
25g
C18H39O4P
MFCD00015351
SFENPMLASUEABX-UHFFFAOYSA-N
trihexyl phosphate
75666
Liquid
Colorless
2528-39-4
[CH3(CH2)5O]3PO
phosphoric acid, trihexyl ester, tri-n-hexyl phosphate, tri n-hexyl phosphate, acmc-20aje0, aronis24109
CCCCCCOP(=O)(OCCCCCC)OCCCCCC
350.48
350.48
≥90%

Chemical Identifiers

2528-39-4
350.48
SFENPMLASUEABX-UHFFFAOYSA-N
75666
CCCCCCOP(=O)(OCCCCCC)OCCCCCC
C18H39O4P
MFCD00015351
phosphoric acid, trihexyl ester, tri-n-hexyl phosphate, tri n-hexyl phosphate, acmc-20aje0, aronis24109
trihexyl phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents