Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-dodecylamine, 95%

$43.73 - $131.15

Chemical Identifiers

Chemical Name or Material Tri-n-dodecylamine
CAS 102-87-4
Molecular Formula [CH3(CH2)11]3N
Formula Weight 522
MDL Number MFCD00008971
Beilstein 1801669
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0447909

 
alfa aesar™
L0447909
10g Each for $43.73

AAL0447918

 
alfa aesar™
L0447918
50g Each for $131.15
Description & Specifications

Specifications

Formula Weight 522
Chemical Name or Material Tri-n-dodecylamine
TSCA TSCA
Assay 95%
Molecular Formula [CH3(CH2)11]3N
MDL Number MFCD00008971
CAS 102-87-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Flash Point 190°C (374°F)
Melting Point 14° to 16°C
Quantity 10g
Beilstein 1801669
Boiling Point 220° to 228°C (0.03mmHg)

Applications
Tri-n-dodecylamine is mainly used in synthesis of cationic and benign surfactants, such as plant detergent, fabric softeners, antistatic agents, anti-corrosive, ore flotation machines and asphalt emulsifiers.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.