Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyltin trifluoromethanesulfonate, tech. 90%

Chemical Identifiers

Chemical Name or Material Tri-n-butyltin trifluoromethanesulfonate
CAS 68725-14-4
Purity Grade Technical Grade
Linear Formula CF3SO3Sn[(CH2)3CH3]3
Formula Weight 439.1
MDL Number MFCD00043324
UN Number UN2923
Beilstein 5818090
Synonym Tri-n-butyltin triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5519203

  SDS
alfa aesar™
H5519203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5519206

  SDS
alfa aesar™
H5519206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55192MD

  SDS
alfa aesar™
H55192MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 439.1
Chemical Name or Material Tri-n-butyltin trifluoromethanesulfonate
Purity Grade Technical Grade
Physical Form Solid
Assay 90%
Linear Formula CF3SO3Sn[(CH2)3CH3]3
MDL Number MFCD00043324
CAS 68725-14-4
UN Number UN2923
Beilstein 5818090
Synonym Tri-n-butyltin triflate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H372-H314-H318-H312
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Sensitivity Moisture Sensitive
Color White to pale brown
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H372-H314-H318-H312
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.