missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyltin trifluoromethanesulfonate, tech. 90%

Manufacturer:  Alfa Aesar™ H5519206

 View more versions of this product

Catalog No. AAH5519206



Specifications

Specifications

Tri-n-butyltin trifluoromethanesulfonate
Moisture sensitive
5g
CF3SO3Sn[(CH2)3CH3]3
UN2923
Tri-n-butyltin triflate
Solid
Technical
113°C (235°F)
Brown to White
68725-14-4
MFCD00043324
5818090
439.1
90%

Chemical Identifiers

68725-14-4
MFCD00043324
Tri-n-butyltin triflate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H314-H318-H312
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H312-H314-H335-H372

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents