missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyltin iodide, tech. 90%, stab. with copper

Manufacturer:  Alfa Aesar™ H5591106

 View more versions of this product

Catalog No. AAH5591106



Specifications

Specifications

Tri-n-butyltin iodide
113°C (235°F)
Light sensitive
Colorless
Stabilized with copper
C12H27ISn
MFCD00074996
bu3sni
CCCC[Sn](CCCC)(CCCC)I
416.962
416.94
90%
1.46 g/mL
1.53
172°C (10mmHg)
5g
7342-47-4
[CH3(CH2)3]3SnI
UN2788
YHHVXVNXTMIXOL-UHFFFAOYSA-M
tributyl(iodo)stannane
23765
Liquid
Technical

Chemical Identifiers

7342-47-4
416.962
YHHVXVNXTMIXOL-UHFFFAOYSA-M
23765
CCCC[Sn](CCCC)(CCCC)I
C12H27ISn
MFCD00074996
bu3sni
tributyl(iodo)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

RTECSNumber : WH8580000

TSCA : No

Recommended Storage : Ambient temperatures

Documents