Please login to your online account to display your discounted pricing

Tri-n-butyltin hydride, 97%, ACROS Organics™

$40.60 - $161.87

Chemical Identifiers

Chemical Name or Material Tri-n-butyltin hydride
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H28Sn
Linear Formula [CH3(CH2)3]3SnH
Formula Weight 291.04
MDL Number MFCD00009416
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC215732500

  SDS
thermo fisher scientific
AC215732500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC215730100

  SDS
thermo fisher scientific
215730100
Glass bottle 10g Each for $40.60

AC215730500

  SDS
thermo fisher scientific
215730500
Glass bottle 50g Each for $161.87
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 291.04
Quantity 250g
Color Clear colorless
Density 1.0800g/mL
Fieser 01,1192; 02,424; 03,294; 04,518; 05,685; 06,604; 07,379; 08,497; 09,476; 10,411; 11,545; 12,516; 14,312; 15,325; 16,343; 17,351
Merck Index 15, 9783
Chemical Name or Material Tri-n-butyltin hydride
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4720 to 1.4740 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C12H28Sn
Linear Formula [CH3(CH2)3]3SnH
MDL Number MFCD00009416
CAS 688-73-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
In contact with water releases f
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Reduction reagent

Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
In contact with water releases f