Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyltin fluoride

Chemical Identifiers

Chemical Name or Material Tri-n-butyltin fluoride
CAS 1983-10-4
Formula Weight 309.04
MDL Number MFCD00013547
UN Number UN3146
Synonym Fluorotri-n-butyltin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3960206

 
alfa aesar™
3960206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3960214

 
alfa aesar™
3960214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.04
Chemical Name or Material Tri-n-butyltin fluoride
TSCA TSCA
Odor Pungent
Physical Form Powder
MDL Number MFCD00013547
CAS 1983-10-4
UN Number UN3146
Synonym Fluorotri-n-butyltin
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Sensitivity Moisture Sensitive
Melting Point 248° to 252°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.