missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyltin bromide, 95%

Manufacturer:  Alfa Aesar™ 7118503

 View more versions of this product

Catalog No. AA7118503



Specifications

Specifications

163°C (12mmHg)
1g
>110°C (230°F)
1.5070
95%
MFCD00010660
Bromotributyltin
CCCC[Sn](CCCC)(CCCC)Br
369.962
369.96
Tri-n-butyltin bromide
1461-23-0
1.338g/mL
Moisture Sensitive
C12H27BrSn
UN2788
FVRKTAOFDKFAMI-UHFFFAOYSA-M
bromo(tributyl)stannane
305767
Liquid

Chemical Identifiers

1461-23-0
369.962
FVRKTAOFDKFAMI-UHFFFAOYSA-M
305767
CCCC[Sn](CCCC)(CCCC)Br
C12H27BrSn
MFCD00010660
Bromotributyltin
bromo(tributyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

RTECSNumber : WH6735000

SDS Documents