Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyltin benzoate, 98%

Chemical Identifiers

Chemical Name or Material Tri-n-butyltin benzoate
CAS 4342-36-3
Linear Formula C6H5CO2Sn[(CH2)3CH3]3
Formula Weight 411.15g/mol
MDL Number MFCD00078009
UN Number UN2788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55449AA

 
alfa aesar™
H55449-AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55449AC

 
alfa aesar™
H55449-AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55449AE

 
alfa aesar™
H55449-AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Refractive Index 1.5180
Density 1.193g/mL
Quantity 5mL
Chemical Name or Material Tri-n-butyltin benzoate
CAS 4342-36-3
Physical Form Liquid
Assay 98%
Linear Formula C6H5CO2Sn[(CH2)3CH3]3
Formula Weight 411.15g/mol
MDL Number MFCD00078009
UN Number UN2788
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER o
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
RTECS Number WH6710000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.