Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyltin acetate, 98%

Chemical Identifiers

Chemical Name or Material Tri-n-butyltin acetate
CAS 56-36-0
Molecular Formula C14H30O2Sn
Formula Weight 349.08
MDL Number MFCD00015228
UN Number UN3146
Synonym Acetoxy(tri-n-butyl)tin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA162550B

 
alfa aesar™
A162550B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA162550I

 
alfa aesar™
A162550I
5000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 349.08
UN Number UN3146
Melting Point 86° to 87°C
Assay 98%
Chemical Name or Material Tri-n-butyltin acetate
Quantity 1000g
Molecular Formula C14H30O2Sn
Health Hazard 2 GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
CAS 56-36-0
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Health Hazard 3 GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Acetoxy(tri-n-butyl)tin
MDL Number MFCD00015228
Safety and Handling

Danger

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.