Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butylphosphine oxide, 97%

Chemical Identifiers

Chemical Name or Material Tri-n-butylphosphine oxide
CAS 814-29-9
Molecular Formula C12H27OP
Formula Weight 218.32
MDL Number MFCD00002082
UN Number UN1759
Beilstein 1761977
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1362906

  SDS
alfa aesar™
A1362906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1362914

  SDS
alfa aesar™
A1362914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.32
Chemical Name or Material Tri-n-butylphosphine oxide
TSCA TSCA
Odor Odorless
Assay 97%
Molecular Formula C12H27OP
MDL Number MFCD00002082
CAS 814-29-9
UN Number UN1759
Beilstein 1761977
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
RTECS Number SZ1575000
Boiling Point 150°C (1mmHg)
Melting Point 64° to 69°C
Quantity 5g

Applications
Tri-n-butylphosphine oxide is used as a catalyst and organic synthesis intermediate. It is also used as a solvent.

Solubility
Soluble in water (56 g/L at 20°C).

Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.