Learn More
Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139341000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tri-n-butylphosphine | |
998-40-3 | |
Colorless to Yellow | |
240.0°C to 242.0°C | |
Authentic | |
Glass bottle | |
1.4610 to 1.4630 | |
MFCD00009462 | |
04, II, 971 | |
0.81 | |
Solubility in water: insoluble. Other solubilities: miscible with ether,methanol,ethanol and benzene | |
CCCCP(CCCC)CCCC | |
202.32 | |
202.32 | |
Liquid |
95% | |
-65.0°C | |
0.8100g/mL | |
37°C | |
94% min. (GC) | |
C12H27P | |
[CH3(CH2)3]3P | |
100 mL | |
01,1191; 05,685; 15,325; 17,269; 06,604; 07,738; 13,158 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
tributylphosphane | |
13831 | |
95% |
Chemical Identifiers
998-40-3 | |
202.32 | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
13831 | |
CCCCP(CCCC)CCCC |
C12H27P | |
MFCD00009462 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
tributylphosphane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long la
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
EINECSNumber : 213-651-2
RTECSNumber : SZ3270000
TSCA : TSCA
RUO – Research Use Only