missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri-n-butylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals

$163.49 - $163.49

Chemical Identifiers

CAS 998-40-3
Molecular Formula C12H27P
Molecular Weight (g/mol) 202.32
MDL Number MFCD00009462
InChI Key TUQOTMZNTHZOKS-UHFFFAOYSA-N
Synonym tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp
PubChem CID 13831
IUPAC Name tributylphosphane
SMILES CCCCP(CCCC)CCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427301000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
427301000
100 mL AcroSeal™ Glass bottle
Each for $163.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

998-40-3
202.32
TUQOTMZNTHZOKS-UHFFFAOYSA-N
13831
CCCCP(CCCC)CCCC
C12H27P
MFCD00009462
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp
tributylphosphane

Specifications

998-40-3
Colorless to Yellow
240.0°C to 242.0°C
Authentic
AcroSeal™ Glass bottle
1.4610 to 1.4630
MFCD00009462
0.81
Solubility in water: insoluble. Other solubilities: miscible with ether,methanol,ethanol and benzene
CCCCP(CCCC)CCCC
202.32
202.32
Liquid
-65.0°C
0.8100g/mL
37°C
94% min. (GC)
C12H27P
[CH3(CH2)3]3P
100 mL
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp
TUQOTMZNTHZOKS-UHFFFAOYSA-N
tributylphosphane
13831
95%
Tri-n-butylphosphine
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long la

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

EINECSNumber : 213-651-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only