missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tri-n-butylphosphine 95%, ACROS Organics™

$63.19 - $255.76

Chemical Identifiers

CAS 998-40-3
Assay Percent Range 95%
Molecular Formula C12H27P
Linear Formula [CH3(CH2)3]3P
Formula Weight 202.32
MDL Number MFCD00009462
Synonym TBUP
Chemical Name or Material Tri-n-butylphosphine, 95%
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC139341000 SDS Thermo Fisher Scientific
139341000
Glass bottle 100mL Each for $63.19
AC139345000 SDS Thermo Fisher Scientific
139345000
Glass bottle 500mL Each for $255.76
Specifications Description & Specifications

Chemical Identifiers

998-40-3
C12H27P
202.32
TBUP
≥94%
95%
[CH3(CH2)3]3P
MFCD00009462
Tri-n-butylphosphine, 95%

Specifications

998-40-3
Glass bottle
Liquid
C12H27P
202.32
0.81g/mL
01,1191; 05,685; 15,325; 17,269; 06,604; 07,738; 13,158
Clear colorless to pale yellow
Tri-n-butylphosphine, 95%
Authentic
1.4610 to 1.4630 (20°C, 589nm)
95%
[CH3(CH2)3]3P
MFCD00009462
04, II, 971
TBUP
100mL
≥94%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-651-2

RTECSNumber : SZ3270000

TSCA : TSCA