Learn More
Alfa Aesar™ Tri-n-butylphosphine, 94%
Manufacturer: Alfa Aesar™ 1411495
Description
- For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels
Specifications
Tri-n-butylphosphine | |
>93° (199°F) | |
1.4620 | |
240°C | |
2 x 500g | |
C12H27P | |
UN3254 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
tributylphosphane | |
13831 | |
Liquid |
0.820g/mL | |
Unpleasant | |
Air & Moisture Sensitive | |
-65°C | |
998-40-3 | |
MFCD00009462 | |
1738261 | |
Insoluble in water; Miscible with ether, methanol, ethanol and benzene | |
CCCCP(CCCC)CCCC | |
202.322 | |
202.32 | |
94% |
Chemical Identifiers
998-40-3 | |
202.322 | |
TUQOTMZNTHZOKS-UHFFFAOYSA-N | |
13831 | |
CCCCP(CCCC)CCCC |
C12H27P | |
MFCD00009462 | |
tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp | |
tributylphosphane |
Safety and Handling
GHS H Statement
H250-H252-H314-H318-H302-H312
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: TRIBUTYLPHOSPHANE
RTECSNumber : SZ3270000
TSCA : TSCA