missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butylphenyltin, 97%

Manufacturer:  Alfa Aesar™ H5559606

 View more versions of this product

Catalog No. AAH5559606



Description

Description

Applications
It is applied in chemical research.

Solubility
Sparingly Soluble (9.9E-5 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications

Specifications

Tri-n-butylphenyltin
113°C (235°F)
125°C to 128°C (0.14mmHg)
5g
C18H32Sn
MFCD00134394
3610571
Sparingly Soluble (9.9E-5 g/L) (25°C)
CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1
367.164
367.14
97%
1.125 g/mL
1.516
Colorless
960-16-7
C6H5Sn[(CH2)3CH3]3
UN2788
phenyl tnbutyl tin
SYUVAXDZVWPKSI-UHFFFAOYSA-N
tributyl(phenyl)stannane
607632
Liquid

Chemical Identifiers

960-16-7
367.164
SYUVAXDZVWPKSI-UHFFFAOYSA-N
607632
CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1
C18H32Sn
MFCD00134394
phenyl tnbutyl tin
tributyl(phenyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P260-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H360FD-H372

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents