Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butylethylphosphonium ethyl sulfate

$142.20 - $142.20

Chemical Identifiers

Chemical Name or Material Tri-n-butylethylphosphonium ethyl sulfate
CAS 654057-98-4
Molecular Formula C14H32P·CH3CH2OSO3
Linear Formula [CH3(CH2)3]3PCH2CH3·CH3CH2OSO3
Formula Weight 356.50g/mol
MDL Number MFCD07368356
Synonym Ethyltri-n-butylphosphonium ethyl sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2786606

 
alfa aesar™
H27866-06
5g Each for $142.20
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Tri-n-butylethylphosphonium ethyl sulfate
CAS 654057-98-4
Molecular Formula C14H32P·CH3CH2OSO3
Linear Formula [CH3(CH2)3]3PCH2CH3·CH3CH2OSO3
Formula Weight 356.50g/mol
MDL Number MFCD07368356
Synonym Ethyltri-n-butylphosphonium ethyl sulfate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.