Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butylborane, 98%

Chemical Identifiers

Chemical Name or Material Tri-n-butylborane
CAS 122-56-5
Formula Weight 182.16
MDL Number MFCD00009423
UN Number UN2845
Synonym Tri-n-butylboron
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3353306

 
alfa aesar™
3353306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3353309

 
alfa aesar™
3353309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3353314

 
alfa aesar™
3353314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.16
Chemical Name or Material Tri-n-butylborane
TSCA TSCA
Refractive Index 1.4285
Physical Form Liquid
Assay 98%
MDL Number MFCD00009423
CAS 122-56-5
UN Number UN2845
Synonym Tri-n-butylboron
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H250-H314-H302
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
RTECS Number ED1850000
Density 0.747g/mL
Flash Point −20° (−4°F)
Sensitivity Air Sensitive
Boiling Point 170°C (222mmHg to 225mmHg)
Melting Point −34°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H314-H302
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Harmful if swallowed.