missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butylamine, 98%

Manufacturer:  Alfa Aesar™ A14868AP

 View more versions of this product

Catalog No. AAA14868AP



Description

Description

Applications
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

Solubility
Miscible with acetone, ethanol, ethyl ether and most organic solvents. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Tri-n-butylamine
Amine-like
<-70°C
10.6
C12H27N
MFCD00009431
1698872
tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7
CCCCN(CCCC)CCCC
185.355
CHEBI:38905
98%
71°C (159°F)
216°C
500mL
102-82-9
[CH3(CH2)3]3N
UN2542
14,9618
IMFACGCPASFAPR-UHFFFAOYSA-N
N,N-dibutylbutan-1-amine
7622
185.36

Chemical Identifiers

102-82-9
185.355
IMFACGCPASFAPR-UHFFFAOYSA-N
7622
N,N-dibutylbutan-1-amine
C12H27N
MFCD00009431
tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7
CHEBI:38905
CCCCN(CCCC)CCCC
Safety and Handling

Safety and Handling

GHS H Statement
H310-H330-H227-H302-H315-H319
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TRIBUTYLAMINE

RTECSNumber : YA0350000

TSCA : TSCA

SDS Documents