Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyl orthoformate, 97%

Chemical Identifiers

Chemical Name or Material Tri-n-butyl orthoformate
CAS 588-43-2
Molecular Formula HC(OCH2CH2CH3)3
Formula Weight 232.36
MDL Number MFCD00015250
Beilstein 1752526
Synonym Orthoformic acid tri-n-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0053906

 
alfa aesar™
L0053906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.36
Chemical Name or Material Tri-n-butyl orthoformate
TSCA TSCA
Assay 97%
Molecular Formula HC(OCH2CH2CH3)3
CAS 588-43-2
Beilstein 1752526
Synonym Orthoformic acid tri-n-butyl ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 247°C
Quantity 5g
MDL Number MFCD00015250
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Flash Point 89°C (192°F)
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.