Learn More
Tri-n-butyl(3-methyl-2-butenyl)tin, 95%, Thermo Scientific Chemicals
$129.75 - $457.03
Chemical Identifiers
CAS | 53911-92-5 |
---|---|
Molecular Formula | C17H36Sn |
Molecular Weight (g/mol) | 359.185 |
MDL Number | MFCD01863649 |
InChI Key | XEFRYQPTIWSVGI-UHFFFAOYSA-N |
Synonym | tributyl 3-methylbut-2-en-1-yl stannane, tributyl 3-methyl-2-butenyl tin, tributylprenylstannane, acmc-1am9p, tri-n-butyl 3-methyl-2-butenyl tin, tributyl 3-methylbut-2-enyl stannane, stannane, tributyl 3-methyl-2-butenyl, tributyl 3-methylbut-2-en-1-yl ;stannane |
PubChem CID | 2734722 |
IUPAC Name | tributyl(3-methylbut-2-enyl)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)CC=C(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2021403
|
Thermo Scientific Chemicals
L2021403 |
1 g |
Each for $129.75
|
|
|||||
AAL2021406
|
Thermo Scientific Chemicals
L2021406 |
5 g |
Each for $457.03
|
|
|||||
Description
Tri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri-n-butyl(3-methyl-2-butenyl)tin is used in prenylation of ethyl propiolate. It is also involved in chemical research.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
53911-92-5 | |
359.185 | |
XEFRYQPTIWSVGI-UHFFFAOYSA-N | |
2734722 | |
CCCC[Sn](CCCC)(CCCC)CC=C(C)C |
C17H36Sn | |
MFCD01863649 | |
tributyl 3-methylbut-2-en-1-yl stannane, tributyl 3-methyl-2-butenyl tin, tributylprenylstannane, acmc-1am9p, tri-n-butyl 3-methyl-2-butenyl tin, tributyl 3-methylbut-2-enyl stannane, stannane, tributyl 3-methyl-2-butenyl, tributyl 3-methylbut-2-en-1-yl ;stannane | |
tributyl(3-methylbut-2-enyl)stannane |
Specifications
53911-92-5 | |
283°C | |
C17H36Sn | |
MFCD01863649 | |
UN2788 | |
tributyl 3-methylbut-2-en-1-yl stannane, tributyl 3-methyl-2-butenyl tin, tributylprenylstannane, acmc-1am9p, tri-n-butyl 3-methyl-2-butenyl tin, tributyl 3-methylbut-2-enyl stannane, stannane, tributyl 3-methyl-2-butenyl, tributyl 3-methylbut-2-en-1-yl ;stannane | |
XEFRYQPTIWSVGI-UHFFFAOYSA-N | |
tributyl(3-methylbut-2-enyl)stannane | |
2734722 | |
95% |
1.042 | |
>110°C (230°F) | |
1.488 | |
1 g | |
3588742 | |
Immiscible with water. | |
CCCC[Sn](CCCC)(CCCC)CC=C(C)C | |
359.185 | |
359.19 | |
Tri-n-butyl(3-methyl-2-butenyl)tin |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only