Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri-n-butyl(1-propenyl)tin, cis + trans, 94%

Chemical Identifiers

Chemical Name or Material Tri-n-butyl(1-propenyl)tin
CAS 105494-65-3
Name Note Cis + Trans
Molecular Formula C15H32Sn
Formula Weight 331.13
MDL Number MFCD03425868
UN Number UN2788
Beilstein 4176362
Synonym 1-Propenyltri-n-butylstannane; 1-(Tri-n-butylstannyl)propene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2020003

  SDS
alfa aesar™
L2020003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 331.13
Chemical Name or Material Tri-n-butyl(1-propenyl)tin
Health Hazard 3 GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Cis + Trans
MDL Number MFCD03425868
CAS 105494-65-3
Recommended Storage 2° to 8°C
Assay 94%
Molecular Formula C15H32Sn
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Boiling Point 77° to 78°C (0.25mmHg)
Quantity 1g
UN Number UN2788
Beilstein 4176362
Synonym 1-Propenyltri-n-butylstannane; 1-(Tri-n-butylstannyl)propene
Safety and Handling

Danger

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C