missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri(ethylene glycol) bis(2-ethylhexanoate), 90+%

$43.88 - $244.30

Chemical Identifiers

CAS 94-28-0
Molecular Formula C22H42O6
Molecular Weight (g/mol) 402.572
MDL Number MFCD00072285
InChI Key FRQDZJMEHSJOPU-UHFFFAOYSA-N
Synonym 2,2'-ethylenedioxydiethyl bis 2-ethylhexanoate
PubChem CID 7185
IUPAC Name 2-[2-[2-(2-ethylhexanoyloxy)ethoxy]ethoxy]ethyl 2-ethylhexanoate
SMILES CCCCC(CC)C(=O)OCCOCCOCCOC(=O)C(CC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54406AE View Documents Alfa Aesar™
H54406AE
100mL Each for $43.88
 
AAH54406K2 View Documents Alfa Aesar™
H54406K2
1L Each for $244.30
 
Description

Description

Applications
It is widely used in water based adhesive, painting and coating industry. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

94-28-0
402.572
FRQDZJMEHSJOPU-UHFFFAOYSA-N
7185
CCCCC(CC)C(=O)OCCOCCOCCOC(=O)C(CC)CCCC
C22H42O6
MFCD00072285
2,2'-ethylenedioxydiethyl bis 2-ethylhexanoate
2-[2-[2-(2-ethylhexanoyloxy)ethoxy]ethoxy]ethyl 2-ethylhexanoate

Specifications

344°C
100mL
C22H42O6
2,2'-ethylenedioxydiethyl bis 2-ethylhexanoate
CCCCC(CC)C(=O)OCCOCCOCCOC(=O)C(CC)CCCC
402.572
402.57
Tri(ethylene glycol) bis(2-ethylhexanoate)
-50°C
94-28-0
MFCD00072285
FRQDZJMEHSJOPU-UHFFFAOYSA-N
2-[2-[2-(2-ethylhexanoyloxy)ethoxy]ethoxy]ethyl 2-ethylhexanoate
7185
≥90%
Safety and Handling

Safety and Handling

RTECSNumber : MO7725000

TSCA : TSCA