missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tri(ethylene glycol) bis(2-ethylhexanoate), 90+%

Manufacturer:  Alfa Aesar™ H54406K2

 View more versions of this product

Catalog No. AAH54406K2



Description

Description

Applications
It is widely used in water based adhesive, painting and coating industry. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
Specifications

Specifications

Tri(ethylene glycol) bis(2-ethylhexanoate)
-50°C
94-28-0
MFCD00072285
FRQDZJMEHSJOPU-UHFFFAOYSA-N
2-[2-[2-(2-ethylhexanoyloxy)ethoxy]ethoxy]ethyl 2-ethylhexanoate
7185
≥90%
344°C
1L
C22H42O6
2,2'-ethylenedioxydiethyl bis 2-ethylhexanoate
CCCCC(CC)C(=O)OCCOCCOCCOC(=O)C(CC)CCCC
402.572
402.57

Chemical Identifiers

94-28-0
402.572
FRQDZJMEHSJOPU-UHFFFAOYSA-N
7185
CCCCC(CC)C(=O)OCCOCCOCCOC(=O)C(CC)CCCC
C22H42O6
MFCD00072285
2,2'-ethylenedioxydiethyl bis 2-ethylhexanoate
2-[2-[2-(2-ethylhexanoyloxy)ethoxy]ethoxy]ethyl 2-ethylhexanoate
Safety and Handling

Safety and Handling

RTECSNumber : MO7725000

TSCA : TSCA

Documents